Spécifications
- Air velocity at the front panel:20 - 4925 ft./min
- Temperature Range:32 to 122 °F (0 to 50 °C)
- Description:Hot Wire Thermoanemometer with NIST-Traceable Calibration
- Air velocity accuracy:±(5% of reading + 1 digit) or ±(1% full scale + 1 digit) (whichever is greater)
- Temperature accuracy:±1.8 °F (±1 °C)
- Cat. no.:76183-370
Spécifications
A propos de cet article
High precision at very low air velocities and flow rates.
- Reliable measurements guaranteed
- Pre-calibrated to NIST-traceable standards
- Measure air velocities up to 4925 ft./min. (25 m/s) to cover a broad range of applications
- Measure airflow up to 99,900 CFM (99,900 CMM)
- Large, dual-function LCD with backlight capability for easy reading
- Auto power-off extends battery life
The Digi-Sense hot-wire thermoanemometer is designed for precision air velocity and airflow measurements in seven user-selectable units (ft/min, m/s, mph, km/h, knots, CFM, or CMM). The instrument will help you meet cooling and environmental requirements for safety and comfort, as well as assure proper ventilation and filtration in cleanrooms and fume hoods.
The large dual display allows you to simultaneously read air velocity or airflow and air temperature and shows real time, maximum, minimum, and average values for the parameters under investigation. Slim telescopic probe is designed to reach into small openings.
The handheld hot-wire anemometer permits single point measurements or average measurements taken over a given time period. Comes in a foam-cut hard carrying case to protect the instrument between uses. Includes software and USB cable to download measurements into a PC for analysis and storage. Low-battery indicator prompts battery replacement, preventing power loss.