Digi-Sense® Pre-calibrated CFM/CMM Vane Thermo Anemometer, Cole-Parmer
# de catalogue 76183-366
Fournisseur: Argos Technologies
Spécifications
- Air velocity at the front panel:80 - 5900 ft./min
- Temperature Range:32 to 122 °F (0 to 50 °C)
- Description:CFM/CMM Vane Thermoanemometer with NIST-Traceable Calibration
- Air velocity accuracy:±(3% + 40 ft./min or 0.2 m/s)
- Temperature accuracy:±4 °F (2 °C)
- Cat. no.:76183-366
- Length:6.75"
- Diameter:2.75"
Spécifications
A propos de cet article
High precision with an extra-large backlit LCD and flow area data storage.
- Reliable measurements guaranteed
- Pre calibrated to NIST-traceable standards
- User-selectable units of measure eliminate the need for conversion
- Measure air velocities up to 5900 ft./min. (30 m/s) to cover a broad range of applications
- Measure air flow up to 999,900 CFM (999,900 CMM)
- Two memory locations, one CFM and one CMM, store commonly used vent area sizes to save time
- Large, dual-function, 4-digit LCD with backlight for easy reading
- Auto power-off extends battery life
- Fits comfortably in the hand
The Digi-Sense CFM/CMM vane thermoanemometer is designed for precision air velocity and airflow measurements in seven user-selectable units of measurement (ft/min, m/s, mph, km/h, knots, CFM, and CMM). The instrument will help you meet cooling and environmental requirements for safety and comfort. The large dual display allows you to simultaneously read air velocity or airflow, and air temperature (°F/°C).
Certifications: NIST-traceable calibration certificate with data, performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.
Delivery: Includes vane probe sensor with 3.9' (1.2 m) cable, three AAA batteries (40 hours typical), and NIST-traceable calibration report with data supplied by InnoCal.
Customers Who Bought This Also Bought