WFI Sampler, Avantor Fluid Handling
Fournisseur: Avantor Fluid Handling
Certificats
A propos de cet article
WFI Samplers are an easy and convenient solution for obtaining perfect fluid samples from a WFI water drop.
- Class VI Compliant Materials: made from AdvantaFlex tubing
- Polypropylene fitting is inert and strong
- Silicone tubing and gasket: unlike TPE materials, silicone will not contribute to TOC counts
- Gamma-irradiation (SAL of 10–6) eliminates additional bio-burden concerns
- Certified free from any animal derived components or processes
Each WFI Sampler is double-polypouched for cleanroom use, and irradiated. The fittings utilize the SterilEnz-II/AT fitting platform, which is pre-gasketed. This means a quick fit to the water system or equipment without further contamination or gasket hassles.
Tubing size: 1/4" I.D. x 7/16" O.D.
Certifications: USP 88 Class VI Biological Test for Plastics. USP 87 MEM Elution Test. USP 661.
Packaging: Individually double-poly pouched.
Customers Who Bought This Also Bought