Whirl-Pak® Round-Bottom Bags for Homogenizer Blenders, Nasco®
# de catalogue 58976-929
Fournisseur: NASCO SAMPLING LLC
Spécifications
- Capacity:1538 ml (52 oz.)
- W×L:185×330 mm (7×13")
- Description:Filter bag Whirl-Pak
- Thickness:4 mil
- Cat. no.:58976-929
- Supplier No.:B01239WA
Spécifications
A propos de cet article
These bags feature a special round bottom seal designed to minimize sample fragments from getting caught in the corners of the bag during blending.
- Specially designed for use with homogenizer blenders
It is made from extra-heavy polyethylene and has special features to make sample testing easier and more efficient. Non-sparking tools should be used for handling.
Customers Who Bought This Also Bought