Isolation Gowns, Dukal
Fournisseur: Dukal
Certificats
A propos de cet article
Isolation gowns are made of high quality spunbonded polypropylene.
- All gowns are made of high quality spunbonded polypropylene
- Fluid-resistant gowns feature a polyethylene coating
- Each gown features elastic cuffs with waist and neck tie closures
- Not made with natural rubber latex
- Universal sizing
- Non sterile
Packaging: These nonsterile gowns are packaged 10 per bag, 5 bags per case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.