Nonsterile Gauze Sponge, DUKAL™ Corporation
Supplier: Dukal
Certificates
About this item
Nonsterile gauze sponges are made of 100% woven cotton.
- Ideal for a number of applications including general cleaning, dressings, prepping, packing, and debriding wounds
- Most non-sterile available in 200 or 100 per bag to suit multiple market needs
- Not made with natural rubber latex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.