Order Entry
ContactUsLinkComponent
COX Colorimetric Inhibitor Screening Assay Kit, Cayman Chemical Company
COX Colorimetric Inhibitor Screening Assay Kit, Cayman Chemical Company
# de catalogue 75817-192
Numéro CAS:  
COX Colorimetric Inhibitor Screening Assay Kit
COX Colorimetric Inhibitor Screening Assay Kit, Cayman Chemical Company
# de catalogue 75817-192
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    192 Wells
  • Cat. no.:
    75817-192
  • Contents:
    Buffer, Heme, COX-1 (ovine), COX-2 (human), Arachidonic Acid (substrate), Potassium Hydroxide, Substrate, Plate, and Cover Sheet
  • Supplier No.:
    701050-192

Spécifications

A propos de cet article

A colorimetric assay for the screening of isozyme-specific inhibitors.

  • Screen for inhibitors of ovine COX-1 or human COX-2
  • Measure the peroxidase component of the COX
  • Assay 41 samples for COX-1 and 41 samples for COX-2 in duplicate
  • Plate-based colorimetric measurement (590 nm)

Cayman's COX Colorimetric Inhibitor Screening Assay measures the peroxidase component of cyclooxygenases. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized N,N,N',N'-tetramethyl-p-phenylenediamine (TMPD) at 590 nm. Inhibition of COX activity, measured by TMPD oxidation, by a variety of selective and nonselective inhibitors, shows changed potencies similar to those observed with other in vitro methods. The COX Colorimetric Inhibitor Screening assay includes both ovine COX-1 and human recombinant COX-2 enzymes in order to screen isozyme-specific inhibitors. Cayman's COX Colorimetric Assay is a time saving tool for screening vast numbers of inhibitors.

Customers Who Bought This Also Bought