Order Entry
ContactUsLinkComponent
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
# de catalogue 75817-124
Numéro CAS:  
Alanine Transaminase Colorimetric Activity Assay Kit
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
# de catalogue 75817-124
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Assay Type:
    Sandwich
  • Reactivity:
    Mouse
  • Size:
    96 Wells
  • Cat. no.:
    75817-124
  • Contents:
    Buffer, Substrate, Cofactor, Initiator, Positive Control, Plate, and Cover Sheet
  • Supplier No.:
    700260-96

Spécifications

A propos de cet article

For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.

  • Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
  • Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
  • Assay Range: 1.56 – 200 ng/mL with an LLOQ of 1.56 ng/mL
  • Rapid assay; get results in under 4 hours
  • LLOD: 0.006 U/mL

Cayman's Ovalbumin IgG1 (mouse) ELISA Kit is an immunometric (i.e., “sandwich”) assay which can be used to measure anti-ovalbumin of the IgG1 isotype in mouse plasma and serum without prior sample purification.

Customers Who Bought This Also Bought