Lattice Support Shelf, Chemglass
# de catalogue 89029-162
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Dimensions:9W×10"D
- Description:Lattice support shelf
- Cat. no.:89029-162
- Supplier No.:CG-3050-10
Spécifications
A propos de cet article
This shelf can be attached to any lattice setup clamp holder.
- Anodized green with a neoprene surface to prevent from slipping off
The ½" square support rod is 7.5" long. Square support rod prevents shelf from rotating once it is attached to the lattice.
Customers Who Bought This Also Bought