A propos de cet article
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP) (ISOBM TD-2 workshop). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: Clone C2
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 75883-794
- 75883-796
- Antigen name:
- AFP
- AFP
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- C2
- C2
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 518808
- Unigene ID: 518808
- Isotype:
- IgG1 kappa
- IgG1 kappa