Magid® EconoWear® Beard Cover, Magid
# de catalogue 69100-516
Fournisseur: Magid Glove
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Clothing Size:One size
- Garment application:Food,Industrial,General Purpose
- Color:White
- Description:Beard Covers
- Fabric:Polypropylene
- Cat. no.:69100-516
- Supplier No.:BC924
Spécifications
A propos de cet article
Disposable white beard covers completely cover beards, mustaches, and sideburns making them ideal for food handling.
- Soft, non-woven, white polypropylene
- Stay on with an elastic strap around the back of the head for comfortable wear
Packaging: 100/dispenser
Customers Who Bought This Also Bought