Reagent Troughs
Fournisseur: TWD Scientific
Certificats
A propos de cet article
Reagent Troughs are made of polypropylene. Troughs support a disposable polypropylene trough liner. Liners are made of thin, disposable, lightweight polypropylene.
- V-shaped bottom and angled sides for maximum reagent recovery
- Usable with up to 12-channel pipettors
- Designed for SBS format applications and compatible with robotic systems
- Reusable
Perfect for 12 or 8-channel pipettes and compatible eith robotic systems.
Customers Who Bought This Also Bought