Graduated Cylinder, Class B, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These volumetric cylinders are made of polypropylene.
They are made up of Class B glasses. Cylinders have ring molded graduations and have blue screening to simplify the measurements.
Certifications: DIN 12681
Customers Who Bought This Also Bought