Tape and Label Dispenser, AK®
# de catalogue 80086-018
Fournisseur: AK
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Tape and label Dispenser
- W×D×H:8.5×6×14"
- Supplier No.:AK-327
Spécifications
A propos de cet article
This dispenser has two removable dowels to accept rolls from 1 to 3" diameter. It works excellent for tapes with peel off backing.
- Made of 3/16" PETG
- Wall mount or free standing
It has a side pocket to keeps pens and markers at fingertips.
Customers Who Bought This Also Bought