ContactUsLinkComponent
Neutrophil/Monocyte Respiratory Burst Assay Kit, Cayman Chemical Company
Neutrophil/Monocyte Respiratory Burst Assay Kit, Cayman Chemical Company
# de catalogue 75817-102
Numéro CAS:  
Neutrophil/Monocyte Respiratory Burst Assay Kit, Cayman Chemical Company
# de catalogue 75817-102
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Neutrophil/Monocyte Respiratory Burst Assay Kit
  • Cat. no.:
    75817-102
  • Supplier No.:
    601130-1
Spécifications
A propos de cet article
Neutrophil/Monocyte Respiratory Burst Assay Kit provides PMA, dihydrorhodamine 123, and additional reagents necessary for inducing and quantifying a respiratory burst response in neutrophils and monocytes by flow cytometry. The assay can be performed on whole blood or on cells in various types of cell culture media. Because the assay reagents are not species-specific, this assay can be used in any species or cell type capable of producing a NADPH oxidase-dependent respiratory burst response.
  • Induce and quantify a respiratory burst response
  • Analyse by flow cytometry
Respiratory burst, or the rapid generation of reactive oxygen species from immune cells, is crucial for the destruction of invading microorganisms by phagocytes. During this process, professional phagocytes convert molecular oxygen to superoxide anions through the action of NADPH oxidase, which is then converted to reactive oxygen species with potent antimicrobial activity.
Delivery: The kit includes dihydrorhodamine 123 assay reagent (50 µL), PMA (1 mM) assay reagent (50 µL), RBC lysis buffer (10X) (10 mL), bovine serum albumin assay reagent (5 g) and calcium chloride (1 M) assay reagent (1 mL).
Customers Who Bought This Also Bought