biOrb® Air Stone
# de catalogue 470234-876
Fournisseur: GEOGLOBAL PARTNERS
Spécifications
- Description:Air Stone
- Cat. no.:470234-876
Spécifications
A propos de cet article
The biOrb® Air Stone fits in the base of the biOrb® underneath the filter cartridge.
- Breaks up the flow of air into the aquarium, splitting it into a mass of fine bubbles
- Reduces noise from the air pump
Over time the tiny pores in the air stone can become blocked by dirt and lime scale preventing air from getting through properly. Depending on your local water supply and the conditions in your biOrb® an air stone can last anywhere from 3 months to 3 years. Changing the air stone from time to time is advisable.
Customers Who Bought This Also Bought