Microscope slide cover glass, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Kemtech america microscope cover glass come in both borosilicate 3.3 and super white glass.
- Individually wrapped boxes
- Excellent optical properties
- Stable Molecular Structures
- Chemically resistant
Packaging: Conveniently packed in 1 oz. hinged boxes; individually vaccum packed in aluminum packaging.
Customers Who Bought This Also Bought