DistriTip® Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
Fournisseur: GILSON, INC.
Certificats
A propos de cet article
The best quality to guarantee repetitive distribution results
DistriTip® tips are polypropylene syringes with easy-to-read level indicators. Three models cover the entire volume range from 1 µl to 1.25 ml. More than 100 aliquots can be dispensed from each syringe. Used with the DISTRIMAN® positive-displacement repetitive pipettor, DistriTip® tips provide more accurate and precise measurements for viscous, dense, volatile, or hot liquids.
DistriTip® Syringe Tips are specifically designed for use with the DISTRIMAN® Repetitive Pipettor.
Customers Who Bought This Also Bought