ContactUsLinkComponent
DISTRIMAN® Repetitive Pipettor, Gilson®
DISTRIMAN® Repetitive Pipettor, Gilson®
Catalog # 76178-538
Supplier:  GILSON, INC.
Product available on GSA Advantage®
DISTRIMAN* Pipette 1 uL to 1.25 mL
DISTRIMAN® Repetitive Pipettor, Gilson®
Catalog # 76178-538
Supplier:  GILSON, INC.
Product available on GSA Advantage®
Specifications
  • Description:
    DISTRIMAN® Repetitive Pipettor
  • Cat. no.:
    76178-538
Specifications
About this item
Adjustable repetetive positive-displacement pipette DISTRIMAN®.
DISTRIMAN® is a positive-displacement, continuously adjustable, repetitive pipette with direct read out designed to simplify multiple dispensing when used with DISTRITIPS® syringes. With DISTRIMAN, you can dispense the exact volume required for any protocol, even fractional volumes. It is ideal for repetitive dispensing in clinical, hospital, biological, chemical, food and beverage, pharmaceutical, cosmetics, and forensics labs.
Customers Who Bought This Also Bought