Hood or Wall Mount Pipette Storage Bins, TrippNT
Fournisseur: TrippNT
Certificats
A propos de cet article
Wall mountable angled large pipette organizers feature five compartments to keep expensive pipettes clean, safe, and readily available.
- Stores expensive volumetric pipettes safely
- Angled bins secure and safely store pipettes
- Provides convenient access to pipettes
- Tape or magnet mount
- White PVC or acrylic construction
- In stock and ready to ship
- American made by TrippNT
- Total replacement Lifetime Guarantee
Installs instantly with tape or magnet. Angled compartments keep pipettes organized with the tip down, while a large opening makes access easy. Completely rust-proof PVC construction. Bright white color gives a clean look and feel to the area.
Customers Who Bought This Also Bought