VWR® Manual Crimping and Decrimping Tools
Fournisseur: VWR International
Certificats
A propos de cet article
Crimping tools provide secure vial closure
- Easy and convenient handling
- Durable, high quality construction
- Crimper size clearly labeled for quick identification
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
Customers Who Bought This Also Bought