Micro Battery-Powered Homogenizer, Omni International
# de catalogue 10015-248
Fournisseur: OMNI INTERNATIONAL
Spécifications
- Description:Micro Battery Powered Homogenizer
- Height:9"
- Sound level:<72 dB
- Power rating:35 W
- Electrical:12 V (lithium ion battery)
- Speed Range:5000 - 30000 rpm
- Volume Range:0.25 – 100 mL
- Weight:466 g
- Supplier No.:UHB-115
- Diameter:3"
Spécifications
A propos de cet article
The Omni Micro Homogenizer is a hand-held battery powered rotor-stator homogenizer designed for use inside or outside of a laboratory setting.
- Variable speed from 5000 to 30000
- Rechargable 12 volt lithium ion battery
- Portable homogenizer
- Compatible with stainless steel and plastic disposable generator probes
The Micro homogenizer is powered by a 12 volt lithium ion battery that provides operating speeds from 5000 to 30000 rpm for rapid homogenization of samples in volumes from 0.20 mL to 100 mL. It is an ideal solution for field based homogenization procedures involving the processing of tissues, fecal material or soft plant samples. When combined with disposable Omni Tip generator probes, cross contamination can be eliminated.
Certifications: CE compliant.
Ordering information: Unit includes 35 watt motor, 12 volt battery, battery charger and user manual. Stainless steel or plastic Omni Tip generator probes sold separately.
Customers Who Bought This Also Bought