Spécifications
- Resolution:0.0001 Abs
- Description:Chemistry Analyzer, with ISE
- Model:BS200/ISE
- Power supply:AC100–130 VAC 200–240 V; 50/60 Hz; 1000 W
- Interfaces:RS 232
- Cat. no.:10023-854
- Supplier No.:BS200/ISE
Spécifications
A propos de cet article
The MedTest 200 Ensemble provides exceptional value and convenience by bringing together low to mid volume laboratory needs to perform clinical chemistry testing.
- Light source provided by halogen-tungsten lamp
- 200 tests per hour/up to 330 tests per hour with ISE
- Bi-directional LIS interface transmission
- Reversed optic system with 8 wavelengths from 340 to 670 nm
Discrete, random access and automated. Automatic probe cleaning, liquid level detection, horizontal and vertical collision protection.