Support Jacks, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
The support jack is resistant to sanitizing agents and autoclavable for sterile applications
- Provides long life, especially when stored and used in corrosive environments
- Can be used to support flasks, jars, columns, large baths, magnetic stirrers, heaters etc.
Support jack deck is constructed of either stainless steel or anodized aluminum, while the lift mechanism for both is constructed entirely from type 304 stainless steel and PTFE.
Customers Who Bought This Also Bought