PREMIRA® II HighDuster Covers, Contec®
Fournisseur: CONTEC, INC. MX
Certificats
A propos de cet article
Disposable Premira II Microfiber HighDuster Covers reduce fiber shedding and provide effective dry and damp dusting for hard to reach areas. The microfiber fabric offers excellent pickup power and is compliant with most cleaning protocols. Compatible with the flexible foam Snake Duster.
- Ideal for hard to reach places
- Easily slips onto a variety of dusting frames
- Disposable, clinically superior alternative to standard dust wands
- Picks up and holds onto debris, rather that spreading it around
- Conformable flexible foam Snake Duster reduces operator strain
Compatible with all QuickConnect™ Handles
Customers Who Bought This Also Bought