Multi-gas/Vapor Smart® Assembled Respirators, Moldex
Fournisseur: Moldex/Metric
Certificats
A propos de cet article
Assembled respirators are ideal for use against multi hazards in the same workplace.
- 100% PVC-Free®
- Use with N95, R95 or P100 particulate filters against even more combinations of contaminants
Application includes degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing, manufacture and use of adhesives, dry cleaning, pesticide or fumigant manufacture and application, treatment of timber and wood products, manufacture and use of inks, printing, manufacture of cosmetic preparations, perfumes and pharmaceuticals, flour bleaching, agriculture, tobacco industry, mining, trucking, manufacture of roofing and asphalt products, petrochemical plants.
Certifications: NIOSH approved for organic vapors, ammonia, methylamine, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen fluoride, formaldehyde, and hydrogen sulfide.
Packaging: Packaged as 1 per bag, 12 bags per case.
Spécifications
- Catalog No:
- 10065-362
- 10065-424
- 10065-426
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health