NORELL™ Select Series™ NMR Tubes, with Caps, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
NORELL™ Select Series™ NMR Tubes are specifically designed for high-resolution NMR.
These tubes are manufactured out of ASTM Type 1 Class A glass as it has a property of high degree of thermal shock resistance and low expansion coefficient. Ideally suitable for variable temperature applications and freeze or thaw cycling.
Customers Who Bought This Also Bought