Conductive Waste Bins, BOTRON COMPANY INC.
Supplier: BOTRON COMPANY INC.
Certificates
About this item
These small ESD safe conductive waste bins are permanently static safe and can be used in manufacturing areas where ESD safety is a concern.
- Made of a conductive polypropylene
- Features a bright yellow ESD logo
Customers Who Bought This Also Bought