VWR® Stirrer Blades with Plain Ends
Supplier: VWR International
Certificates
About this item
Blades fit PTFE stirring shaft; hole diameter 6.5 mm, 3.2 mm thick.
- For plain shafts
- Very good chemical resistance
- Inert
- Temperature resistant up to +280°C
Ordering information: For use with 005.500 series plain stirrer shafts (sold separately). Contact your Avantor representative for more information.
Customers Who Bought This Also Bought