White Swan Meta Unisex Consultation Coat, Encompass
Fournisseur: White Swan
Certificats
A propos de cet article
These white Unisex 30" Consultation Lab coats are manufactured from 65% polyester and 35% cotton.
- Seven pockets with two inside
- Two oversized lower pockets to hold iPad
- Each outside lower patch pocket has concealed inside pocket
- Fine line twill with soil release
- Pen divider on chest pocket
Spécifications
- Catalog No:
- 20030-606
- 20030-604
- 20030-616
- 20030-610
- 20030-608
- 76103-176
- 20030-614
- 20030-612
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton