ActivArmr® 97-505 Gloves, Ansell
Fournisseur: Ansell Canada
Certificats
A propos de cet article
Designed to deliver comfort, barehand dexterity, and excellent cut resistance.
- At least 8 times more cut protection than most popular leather and construction gloves
- Featuring Stretch Armor Yarn – a unique combination of DuPont™ Kevlar® and Ansell Intercept Technology®
- Foam nitrile coating delivers great wear and grip
- Material: DuPont™ Kevlar® and Ansell Intercept Technology®
ActivArmr® 97-505 gloves have been designed with Intercept yarn and DuPont Kevlar to provide excellent cut resistance, comfort, and dexterity. The foam nitrile coating on this glove gives wearers the ability to handle small obejcts and provides excellent wet and dry grip. 97-505 is ergonomically designed to provide a naturally curved, relaxed fit to reduce hand fatigue.
Certifications: Gloves offer ANSI Level 5 Abrasion and ANSI A4 cut resistance.
Spécifications
- Catalog No:
- 75982-616
- 75982-618
- 75982-620
- 75982-622
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Palm Coated
- Palm Coated
- Palm Coated
- Palm Coated
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist