Metal Closed Head Pail, Qorpak®
# de catalogue 75880-448
Fournisseur: Qorpak
Spécifications
- Cap Size:2"
- Capacity:5 gal. (640 oz.)
- Color:Black
- Material:Metal
- Supplier No.:MET-03035
Spécifications
A propos de cet article
24-gauge black steel phenolic lined closed head pail with 2" bung and Rieke plug.
Customers Who Bought This Also Bought