VWR® Septa Sealing Mat
# de catalogue 470166-590
Fournisseur: VWR International
Spécifications
- Description:Septa Sealing Mat
Spécifications
A propos de cet article
This 96-well septa sealing mat is a cost-effective alternative and is compatible with the ABI 3100.
- 96-well sealing mat with septa
- Round well
- Non-perforated
The septa sealing mat is compatible for use on the ABI 3100. Standard packaging includes 10 mats/pack, 5 packs/case. In addition to the standard packaging option, it is also available in packs of 10.
Customers Who Bought This Also Bought