No. 1 Rectangular Cover Glass, Azer Scientific
# de catalogue 10862-522
Fournisseur: Azer Scientific Inc.
Spécifications
- Numbering:#1
- Description:Cover glasses, rectangular
- L×W:50×24 mm
- Cat. no.:10862-522
- Packaging:2 Oz./Pack, 10 Packs/Case
- Supplier No.:112450-20
Spécifications
A propos de cet article
No
- Ultra-flat and chemical-resistant
- Does not emit disturbing fluorescence
- Appropriate for use in cover slippers
Azer Scientific's cover glass is made of pure white borosilicate glass of the finest hydrolytic class. The ultra-flat, chemical-resistant surface offers optimal manual and automated cover slipping. Additionally, it will not emit disturbing fluorescence under the UV light of the fluorescence microscopy. The popular 2 oz. packaging saves the user additional shipping fees and space.
1 cover glass.
Customers Who Bought This Also Bought