Spécifications
- Capacity:20 ml
- Sterility:Non sterile
- Description:Screw Top Pathology Container
- Cat. no.:95041-504
- Supplier No.:PFNBF-20-EML
Spécifications
A propos de cet article
This 20 mL pathology container is supplied with lid.
- Polypropylene containers with polyethylene screw caps
- 95 kPa compliant
This non sterile pathology container is 95 kPa compliant and comes with a screw cap lid for closure. Two clicks ensure the seal is fully engaged for safe collection and transport of the specimen.