Wallcur® PRACTI-Carpuject
# de catalogue 470232-758
Fournisseur: WALLCUR, LLC.
Spécifications
- Description:PRACTI-Carpuject Holders
- Cat. no.:470232-758
Spécifications
A propos de cet article
Perfect For Training
- Learn How To Prepare And Administer Specific Dosages
- Accepts Standard Cartridges
Reusable plastic cartridge holders are available in packages of five. Designed to provide both stability during injection and easy, safe disposal of the glass cartridge unit after use.
Delivery: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Customers Who Bought This Also Bought