Wallcur® PRACTI-Bacteriostatic Water
# de catalogue 470232-744
Fournisseur: WALLCUR, LLC.
Spécifications
- Description:PRACTI-Bacteriostatic Water
- Cat. no.:470232-744
- Includes:Distilled Water
Spécifications
A propos de cet article
Great For Your ER And EMS Training Lab
- Contains No Medication
- Use As A Dilatant For Reconstitution
- Pre-Filled Syringe
PRACTI-Bacteriostatic Water™ is a 1 mL pre-filled syringe containing distilled water. Wallcur's PRACTI-Bacteriostatic Water simulates a diluent that can be used for reconstitution training. An important training aid for your nursing skills lab, EMT field training simulations, and more.
Delivery: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Customers Who Bought This Also Bought