Wallcur® PRACTI-Ampule Safety Breakers
Fournisseur: WALLCUR, LLC.
Certificats
A propos de cet article
Safe And Disposable
- Contains No Medication
- Available In 2 Sizes
- Designed For Safe And Accurate Opening Of Ampules
- After Use, Dispose Of Broken Ampule And Breaker
Increase student confidence and competency during ampule handling and practice with these easy to use ampule breakers. Students will avoid direct finger contact with broken glass during and after practice thus eliminating the fear of cuts from poor technique. These safe, reliable and economical Ampule Breakers make ampule manipulation much easier and ampule breakage at the neck quicker and cleaner. Dispose of ampule and breaker together in a proper sharps container following practice. Available in 2 sizes to fit the WALLCUR PRACTI-Ampules.
Delivery: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Customers Who Bought This Also Bought