White Polypropylene Screw Caps, Pulp/Vinyl Liner,Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Polypropylene caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability.
- Perfect for general use
- Not recommended for active hydrocarbons or bleaches
Pulp/Vinyl Lined Polypropylene Caps are commonly used for food, beverage, medical and chemical applications packed at less than 120 °F., including mild acids, alkalis, solvents, alcohols, oils, and aqueous products.
Customers Who Bought This Also Bought