Straight Sided Rounds, Amber, Wide Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Amber Straight Sided Round Glass Jars are designed without shoulders for maximum storage capacity and UV protection.
- Minimal chemical interference with contents
- Amber glass jars absorb all wavelengths in the ultraviolet spectrum
- Ensure integrity for long term storage
- Offer excellent protection for UV-sensitive contents
Also known as Paragon Jars, these multi-purpose jars are very versatile.
Customers Who Bought This Also Bought