A propos de cet article
The lightest weight cut resistant and oil repellent glove providing enhanced worker comfort and safety, with strategically reinforced 'high wear' regions for extended-use life. These gloves are made up of Dyneema® diamond technology material.
- RIPEL™ oil repellent technology for maintaining dry hands in oily environments
- Thumb crotch reinforcement technology for up to 12× the durability of the inherently weak thumb crotch region*
- ANSELL GRIP™ Technology provides an average of 13% better grip performance for safe handling of Dry and Oily items**
The HyFlex® 11-937 is an ultra-lightweight, cut resistant and oil repellent ³/₄ coated glove with reinforced thumb crotch that provides up to 12× the durability for extended wear.
Designed to feature a full range of Ansell Product Technologies designed for maximum worker and product protection. RIPEL™ Technology prevents liquids and oil from making skin contact keeping hands safe and dry. ANSELL GRIP™ Technology delivers superior grip performance for safer handling of wet or oily parts. FORTIX™ Technology provides extended glove life and up to high abrasion resistance. ZONZ™ Comfort Fit Technology provides dexterity and comfort.
*compared to unreinforced TCR competitor gloves
**compared to competitive set 2017
18 gauge liner featuring Dyneema® Diamond Technology provides high dexterity and comfort. The nitrile coating and integrated thumb crotch reinforcement (TCR) offer long-lasting protection and 12× better wear life than unreinforced gloves based on field tests. ³/₄ coated dip provides liquid repellence. A silicone free design insures full product protection.
Spécifications
- Catalog No:
- 75927-994
- 75927-996
- 75927-998
- 75928-000
- 75927-846
- 75927-848
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Palm Fit
- Palm Fit
- Palm Fit
- Palm Fit
- Palm Fit
- Palm Fit
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18