Somso® Arthroscopic Wrist
# de catalogue 470233-288
Fournisseur: MARCUS SOMMER
Spécifications
- Description:Arthroscopic Wrist
- Cat. no.:470233-288
Spécifications
A propos de cet article
Highly Detailed.
- Removable bones and joints
- 2 pre built access points
- Replacable parts
According to PD Dr. med. H. Hempfling of Murnau. The model consists of a plastic hand in which the carpal bones, the radius and ulna, together with the carpal disc and the intra-articular ligaments are visible. On the extensor sideof the hand , there are two points of access to the inner cavity of the joint: a radiodorsal and an ulnodorsal opening. The carpal disc can be attached to the ulna and the carpal ligaments on both the flexor and extensor sides, and can be exchanged or replaced as required.
Customers Who Bought This Also Bought