Osmometer, Advanced Instruments | Avantor
Osmometer, Advanced Instruments
Single-Sample Micro-Osmometer, 20 µL Single Sample
Catalog # 76193-112
Fournisseur #:  OSMO1
Osmometer, Advanced Instruments
Catalog # 76193-112
Osmometer, Advanced Instruments
Osmometer, Advanced Instruments
Single-Sample Micro-Osmometer, 20 µL Single Sample
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each
Product Details & Documents
Best in class single-sample micro-osmometer that is easy-to-use offers precise, accurate results.
  • Industry leading accuracy and precision
  • One step direct sampling
  • Easy-to-use, intuitive touchscreen user interface
  • Small 20 µL sample size
  • Secure and efficient data management
  • Built in quality control
High-quality, easy-to-use single-sample micro-osmometer for those laboratories who prefer to directly draw and test small sample volumes and want accurate, precise results and the security and efficiency of data management.
Certifications: CE. ETL. RoHS. IVD.
Ordering information: General supplies (sold separately) include Micro-Sample Test Kit [133800], Printer paper (5 rolls) [FLA835], 20 µL East-Eject™ Sampler [3M0825], Replacement sample probe [133060R], 50 mOsm/kg Calibration Standard [3MA005], 850 mOsm/kg Calibration Standard [3MA085], and 2000 mOsm/kg Calibration Standard [3MA200].
Spécifications
  • Désignation:
    Single-Sample Micro-Osmometer, 20 µL Single Sample
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Osmometer Sample Tips and Cleaners (500 each) with One Plunger Wire
    Osmometer Sample Tips and Cleaners (500 each) with One Plunger Wire
    76193-110
    Log in pour la disponibilité
    Each - $4,433.00