ActivArmr® 43-217 TIG Welding Gloves, Ansell
Supplier: Avantor
Certificates
About this item
TIG welding glove for precise welding and thermal jobs.
- Provides protection from heat, flames, sparks, and weld spatter
- Premium goatskin for a relaxed, natural feel
- Aramid stitching for improved durability and cost of ownership
- Ideal for precise, technical welding applications
ActivArmr® 43-217 is a TIG welding glove that provides EN 12477 Type-B protection for precise welding and thermal tasks. It is suitable for TIG welding, as well as grinding, brazing, metal fabrication, and maintenance. These gloves provide protection from heat, flames, sparks, and weld spatter. The three-dimensional thumb design greatly increases fine dexterity for precise welding control. Premium full grain goatskin and welted seams with para-aramid thread ensure durability in use. The long-split cowhide cuff extends protection to the forearms.
ActivArmr® 43-217 provides comfort, dexterity, and protection in a cost effective thermal glove.
Specifications
- Catalog No:
- 75840-576
- Ambidextrous/Hand-specific:
- Hand-specific
- Disposable / reusable:
- Reusable
- Latex/Latex-free:
- Latex
- Powdered/Powder-free:
- Powder-free
- Cuff style:
- Gauntlet