ActivArmr® Workguard™ 43-216 Leather Welding Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Heavy-duty welding glove for demanding welding and thermal job tasks.
- Provides EN 12477 Type-A protection
- Suitable for stick and MIG welding, as well as torch cutting and grinding
- Comfortable cotton lining provides thermal insulation and extended comfort
- Long cuff extends protection over the forearm
ActivArmr® 43-216 has an improved three-dimensional thumb design that greatly increases fine dexterity for precise welding control. Its premium split cowhide leather and welted seams with Kevlar® thread ensure durability in use while providing comfort and protection for the wearer.
High levels of durability, control, and protection from heat, flame, sparks, and puncture.
Specifications
- Catalog No:
- 75796-494
- 75796-498
- 75796-496
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Kevlar®/Cotton
- Kevlar®/Cotton
- Kevlar®/Cotton
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet