SP Wilmad-LabGlass Benchtop Spectrometer NMR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Ideal for use with 43, 60, or 80 MHz manual sample loading benchtop NMR spectrometers, Wilmad-LabGlass Benchtop NMR tubes have been tested and approved by multiple benchtop spectrometer manufacturers, providing peace of mind and confidence when purchasing consumables for the instrument.
- Economically priced for a high-throughput laboratory environment
- 5 mm Ext.Ø tubes available in 7 or 8" lengths, in quantities of 25 or 150 pieces
- Packaging allows for easy tube access and storage
- Type 1, Class B Borosilicate glass construction with disposable caps
Caution: Not for use with high-field instruments or spinning experiments.
Customers Who Bought This Also Bought