CurTec HDPE Wide Neck Drums With Red PP Lids, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
This High Performance Packaging line from CurTec provides the highest level of product protection and added value through quality, functionality, ease of use, and design.
- Water tight closure
- Tamper-evident feature
- Stable stacking through a clever base and lid construction
- UN marking for solids
Applications include shipping and stocking valuable fine chemicals, API’s, and intermediates, as well as finished products in powder, tablet, and paste format. The line is backed by a quality assurance program which includes HACCP, ISO certification, and a Drug Master File for all packaging products.
Customers Who Bought This Also Bought