Microflex® 93-850 Ultimate Barrier Disposable Nitrile Gloves, Ansell
Supplier: Microflex
Certificates
About this item
Ultimate barrier disposable nitrile glove with TNT™ technology.
- 2X more chemical splash protection than leading brands due to breakthrough polymer bonding technique
- Exceeds every known standard for barrier quality and consistency (Low 0.40 AQL)
- Made with TNT™ Chemical Splash resistance technology, for soft durable protection against a wide range of chemicals
- Silicone-free formulation for product protection
Microflex® 93-850 nitrile disposable gloves represent the ultimate barrier against harmful exposures. These tough, durable gloves stand up to tough jobs while significantly reducing workers’ risk of exposure to hazardous materials.
Caution: Based on EN 374 and ASTM F739 chemical permeation test data when compared to performance of disposable nitrile gloves of a similar weight. Based on Ansell durability pull test data, when compared to performance of disposable nitrile gloves of similar thickness. Microflex® 93-850 0.40 AQL is lower than the AQL of other known industrial and medical grade disposable gloves.
Specifications
- Catalog No:
- 76169-806
- 76169-808
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Cuff length:
- 9.5"
- 9.5"
- Finger thickness:
- 7.5 mil
- 7.5 mil
- Palm thickness:
- 4.7 mil
- 4.7 mil
- Ultimate elongation:
- 550%
- 550%