Cleanert® CN SPE Plates, 96-Well, Bonna Agela
Fournisseur: Agela Technologies
Certificats
A propos de cet article
These 96-well plates have a particle size of 40 to 60 µm and a pore size of 60 Å.
Customers Who Bought This Also Bought
These 96-well plates have a particle size of 40 to 60 µm and a pore size of 60 Å.