Vydac 302IC, Anion Exclusion HPLC Column
Fournisseur: Avantor
Certificats
A propos de cet article
Vydac 302IC is a low capacity anion-exchange (quaternary amine) material based on a high purity 10 µm large pore size silica.
- Used with standard HPLC systems for analysis of common ions
- Widely used for environmental analysis
This column is well known for 'non-suppressed' ion chromatography applications.
Vydac is a registered trademark of W.R.Grace and Co.
Customers Who Bought This Also Bought